Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
N,N-Dimethylformamid

N,N-Dimethylformamid

2865, Glass bottle, 1.000 ml
Catalog # SCARDI10731000
Flammable
Health
Irritant
Supplier:   Scharlau
N,N-Dimethylformamid
Catalog # SCARDI10731000
Supplier:  Scharlau
CAS Number:  68-12-2
N,N-Dimethylformamid
N,N-Dimethylformamid

2865, Glass bottle, 1.000 ml

CAS Number:  68-12-2
Price
Inquire For Price
Log in for availability
1 * 1.000 mL
Product Details & Documents
Synonyms:  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09
  • Merck Index:13,03269
  • Boiling Pt:153 °C (1013 hPa)
  • Melting Pt:–61 °C
  • Density:0,949 g/cm³ (20 °C)
  • Flash Pt:57,5 °C (ot. pohárek)
  • UN:2265
  • ADR:3,III
  •  :Density: 0,949 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-679-5
  • REACH:01-2119475605-32
Specifications

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    1.000 ml
  • Sekvence:
    2865
  • Cat. No.:
    SCARDI10731000

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.