Orders LinkContactUsLinkComponent
Pipe Marker, Steam
Pipe Marker, Steam
Pipe marker, steam, Polyester, 150×12 mm
Catalog # BRDYN005604
Supplier:   Brady
Pipe Marker, Steam
Catalog # BRDYN005604
Supplier:  Brady
Pipe Marker, Steam
Pipe Marker, Steam
Pipe marker, steam, Polyester, 150×12 mm
Price
405,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Individual pipe markers on map with perforated arrows, without pictogram.
  • Polyester with polyester overlaminate
  • Permanent rubber-based adhesive
  • Coloir: Black on grey
 : Total quantity per pack - five marker(s)/card.
Specifications
  • Popis:
    Pipe marker, steam
  • Rozměry:
    150×12 mm
  • Materiál:
    Polyester