Orders LinkContactUsLinkComponent
Pipe Marker, Compressed Air
Pipe marker, compressed air, Polyester, 150×12 mm
  BRDYN004705
 :   Brady
Pipe Marker, Compressed Air
  BRDYN004705
 :  Brady
Pipe Marker, Compressed Air
Pipe Marker, Compressed Air
Pipe marker, compressed air, Polyester, 150×12 mm
405,00 Kč
1 * 1 KS
Individual pipe markers on map with perforated arrows, without pictogram.
  • Material: Self-adhesive laminated polyester
  • Colour: White on blue
  • Weatherproof; UV and chemical resistant
Available in multiple sizes and on or without a liner.
  • Popis:
    Pipe marker, compressed air
  • Rozměry:
    150×12 mm
  • Materiál:
    Polyester