Orders LinkContactUsLinkComponent
Face Shield Plus
Face Shield Plus
Face shield plus
Katalogové číslo PRTWPW96CLR
Dodavatel:   Portwest
Face Shield Plus
Katalogové číslo PRTWPW96CLR
Dodavatel:  Portwest
Face Shield Plus
Face shield plus
Přejít na:
Price
641,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
This face shield protects against molten metal splashes and hot solid projections.
  • Browguard including clear polycarbonate visor with aluminum edge
  • Protects from impacts, metal splashes, and hot solid projections
  • Suitable for a wide range of applications
  • Wheel ratchet size adjustment for easy fitting
  • Simple and secure visor replacement
  • Retail bag aids presentation for retail sales
Superior front and side face protection. Ratchet adjustment for comfort fit. Replacement visor available (PW99).
Certifications: ANSI/ISEA Z87 +.
Specifikace
  • Popis:
    Face shield plus