0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Enzymy
Pepsin
Pepsin
Catalog # SIAL77161-100G
Supplier:
Merck
CAS Number:
Expand image
Pepsin
Catalog # SIAL77161-100G
Supplier:
Merck
CAS Number:
Specifications
Typ balení:
Glass bottle for solids
Bal.:
100 g
Čí. CAS:
9001-75-6
Podmínky uskladnění:
Refrigerator
Název enzymu:
Pepsin
Specifications
About this item
Product Details
Product Details