Order Entry
Czech Republic
ContactUsLinkComponent
Diethylenglykolmonobutylether, Sigma-Aldrich®
Irritant
Diethylenglykolmonobutylether, Sigma-Aldrich®
Catalog # SIAL32250-1L-F
Supplier:  Merck
CAS Number:  
Irritant
Diethylenglykolmonobutylether, Sigma-Aldrich®
Catalog # SIAL32250-1L-F
Supplier:  Merck
CAS Number:  

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    1 L
  • Sekvence:
    5037

Specifications

About this item

 :
Merck Index:13,01556
Boiling Pt:230,4 °C (1013 hPa)
Melting Pt:–68 °C
Density:0,954 g/cm³ (20 °C)
Flash Pt:100 °C
Storage Temperature:Ambient
EINECS:203-961-6
 :GHS07
CAS Number:112-34-5
Formula:CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MDL Number:MFCD00002881