2-Bromo-5-(trifluoromethyl)pyridine
Catalog # ABCRF01598G
Supplier: abcr
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:5 g
- Sekvence:3173
Specifications
About this item
:
CAS Number:50488-42-1
Formula:C₆H₃BrF₃N
MDL Number:MFCD00153086