Orders LinkContactUsLinkComponent
Avantor uvádí vědu do pohybu pro lepší svět | Avantor
Dodavatel:   Deb-STOKO
Dodavatel:  Deb-STOKO
Certifikáty
2 Dostupných variant níže
Product Details & Documents
This white dispenser is designed for applications in general industry, education, hospitality and more.
  • Wall-mounted
  • Durable plastic
  • One hand operation
BioCote® Protected - These dispenser buttons are exclusively protected with BioCote® - a silver-based antimicrobial agent that is incorporated into plastic components at the time of manufacture and inhibits the growth of a broad spectrum of bacteria and molds. BioCote® will not wear out or rub off and is effective for the lifetime of the dispenser button.
Product Family Options
Informace o produktu
  • Sekvence
    Barva
    Kapacita
    Š×H×V
    Dostupnost
    Price
  • EVONSHW1LDS-UK
    Clense shower dispenser
    Clense shower dispenser
    02
    Bílá
    1 L
    132×117×235 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 785,00 Kč
    785,00 Kč
    1 * 1 KS
       785,00 Kč
  • EVONLGT1LDS-UK
    Clense light dispenser
    Clense light dispenser
    01
    Bílá
    1 L
    132×117×235 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 785,00 Kč
    785,00 Kč
    1 * 1 KS
       785,00 Kč