Conductivity Sensor, Memosens CLS15E
Catalog # ENDRCLS15E-1061/0
Supplier: Endress+Hauser
Specifications
- Připojení:Thread NPT1/2
- Popis:Conductivity sensor, Memosens CLS15E, non hazardous area approval
- Bal.:1
- Konstanta cely:K=0,01
Specifications
About this item
A digital sensor for low measuring ranges, suitable for applications such as boiler feed water monitoring and chip cleaning. Benefit from its extended data storage functions that enable predictive maintenance.
- Designed for durability, the sensor requires minimal maintenance and is characterized by a long service life
- Its sterilisable and autoclavable properties allow installation in sterile environments within plants
- With its electrode geometry, the Memosens CLS15E provides accurate readings even at low conductivities, ensuring reliability
- A certificate ensuring the specific cell constant allows for exact fine-tuning of the measuring point, ensuring precision in adjustment
The Memosens CLS15E is the ideal choice for measuring conductivity in environments with low measuring ranges, such as boiler feed water and chip cleaning. This contact-based conductivity sensor ensures reliable and accurate performance, even in hazardous environments. With its Memosens 2.0 digital technology, the CLS15E has an extended memory for calibration, setting and process data, enabling predictive maintenance and providing an excellent basis for IIoT services.
Delivery: Digital conductivity sensor Memosens CLS15E.