Orders LinkContactUsLinkComponent
Conductivity Sensor, Smartec CLD18
Conductivity Sensor, Smartec CLD18
Conductivity sensor, Smartec CLD18, stainless steel housing, IP 69, NEMA6P (body), Tri-clamp ISO 2852 50,8 mm
Katalogové číslo ENDRCLD18-1014/0
Dodavatel:   Endress+Hauser
Conductivity Sensor, Smartec CLD18
Katalogové číslo ENDRCLD18-1014/0
Dodavatel:  Endress+Hauser
Conductivity Sensor, Smartec CLD18
Conductivity Sensor, Smartec CLD18
Conductivity sensor, Smartec CLD18, stainless steel housing, IP 69, NEMA6P (body), Tri-clamp ISO 2852 50,8 mm
Přejít na:
Price
61 740,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Compact conductivity system for hygienic applications.
  • The compact design allows installation in narrow pipes without obstructing the flow
  • Effortlessly integrates into your plant infrastructure with the optional IO-Link protocol
  • The product's unique hygienic design prevents contamination, has 3-A approval and complies with EG regulations 2023/2006 and 1935/2004
  • Designed for cleaning-in-place (CIP)
Ordering information: For optional items or more information, contact your Avantor representative.
Delivery information: CLD18 compact conductivity system.
Specifikace
  • Připojení:
    Tri-clamp ISO 2852 50,8 mm
  • Popis:
    Conductivity sensor, Smartec CLD18, stainless steel housing, IP 69, NEMA6P (body)