0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Domů
iFluor®
iFluor® 790, Styramide *Superior Replacement for Alexa Fluor 790 tyramide*
iFluor® 790, Styramide *Superior Replacement for Alexa Fluor 790 tyramide*
iFluor® 790, Styramide *Superior Replacement for Alexa Fluor 790 tyramide*
Catalog # AATB45070
Supplier:
AAT Bioquest
CAS Number:
Expand image
iFluor® 790, Styramide *Superior Replacement for Alexa Fluor 790 tyramide*
Catalog # AATB45070
Supplier:
AAT Bioquest
CAS Number:
Specifications
Bal.:
100
Sekvence:
0100
Specifications
About this item
Product Details
:
Product Details