ReadiUse™ Preactivated HRP NHS Ester
Katalogové číslo AATB11025
Dodavatel: AAT BIOQUEST
Číslo CAS:
Specifikace
- Bal.:300 µG
- Popis:ReadiUse™ preactivated HRP NHS ester
Specifikace
O tomto produktu
ReadiUse™ Preactivated HRP NHS ester is the mono NHS ester of HRP.
- Ready-to-use
- Highly useful antibody label
- Widely used for immunoassays
- Multiple applications
Caution: For research use only.