ContactUsLinkComponent
ddGTP [2',3'-Dideoxyguanosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
ddGTP [2',3'-Dideoxyguanosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Catalog # AATB17210
Supplier:  AAT Bioquest
CAS Number:  
undefined
ddGTP [2',3'-Dideoxyguanosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Catalog # AATB17210
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Bal.:
    1 µmol
  • Popis:
    ddGTP [2',3'-dideoxyguanosine-5'-triphosphate] *10 mm in ddH₂O*
Specifications
About this item
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Caution: For research use only.