Amplite® Fluorimetric Fluorescamine Protein Quantitation Kit *Blue Fluorescence*
Catalog # AATB11100
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Bal.:200 Tests
- Popis:Amplite™ fluorimetric fluorescamine protein quantitation kit *blue fluorescence*
- Testů v kitu:200 Tests
Specifications
About this item
Fluorescamine is intrinsically non-fluorescent but reacts rapidly with primary aliphatic amines, including those in peptides and proteins, to yield a blue-green-fluorescent derivative.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.