Amplite® Colorimetric Biotin Quantitation Kit
Catalog # AATB5522
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Bal.:200 Tests
- Popis:Amplite™ colorimetric biotin quantitation kit
- Testů v kitu:200 Tests
Specifications
About this item
The avidin/streptavidin-biotin interaction is the strongest known non-covalent biological interaction (Kd = 10 to 15 M⁻¹) between a protein and its ligand.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.