StrandBrite™ Green Fluorimetric RNA Quantitation Kit *High Selectivity*
Catalog # AATB17657
Supplier: AAT Bioquest
CAS Number:
Specifications
- Bal.:100 Tests
- Popis:StrandBrite™ green fluorimetric RNA quantitation kit *high selectivity*
Specifications
About this item
The major challenge to analyse RNA in live cells is the interferences caused by DNA.
- Highly sensitive RNA detection
- Robust protocol
- Multiple applications
Caution: For research use only.