Cell Navigator® Lysosome Staining Kit *Green Fluorescence with 405 nm Excitation*
Catalog # AATB22651
Supplier: AAT Bioquest
CAS Number:
Specifications
- Bal.:500 Tests
- Popis:Cell Navigator™ lysosome staining kit *green fluorescence with 405 nm excitation*
Specifications
About this item
Our Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labelling sub-cellular organelles such as membranes, lysosomes, mitochondria and nuclei etc.
- Cell-permeant staining
- Improved signal
- Suitable for all major fluorescence platforms
Caution: For research use only.