Native Amanita phalloides Phalloidin (from Amanita phalloides), Alexa Fluor® 680
Catalog # AATB23128
Supplier: AAT Bioquest
CAS Number:
Specifications
- Typ balení:Vial
- Bal.:300 Tests
- Spojení:Alexa Fluor® 680
- Typ proteinu/peptidu:Native
- Zdroj:Amanita phalloides
- Druhy:Amanita phalloides
- Proteinová synonyma:heptapeptide phallotoxin
- Název proteinu/peptidu:Phalloidin
Specifications
About this item
This NIR fluorescent phalloidin conjugate selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.
- Brighter signal
- High affinity and selectivity
- In house quality control
Caution: For research use only.