Portelite™ Fluorimetric Total Nucleic Acid Quantitation Kit
Catalog # AATB17631
Dodavatel: AAT BIOQUEST
Číslo CAS:
Specifikace
- Bal.:200 Tests
- Popis:Kit is designed to rapidly measure the total amounts of nucleic acids, including double-stranded DNA (dsDNA), single-stranded DNA (ssDNA) and RNA in a sample
Specifikace
O tomto produktu
Portelite™ fluorimetric total nucleic acid quantitation kit is optimised for measuring the total amounts of nucleic acids with CytoCite™ or Qubit® fluorometers.
- Optimized for cytocite and qubit fluorometers
- Sensitive and indiscriminate binding
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer, and pre-diluted DNA standards.
Packaging: Plastic vial.
Caution: For research use only.