Helixyte™ Green Fluorometric dsDNA Quantitation kit *High Sensitivity*
Catalog # AATB17651
Supplier: AAT Bioquest
CAS Number:
Specifications
- Bal.:200 Tests
- Popis:Helixyte™ green fluorimetric dsDNA quantitation kit *high sensitivity*
Specifications
About this item
Helixyte™ Green dsDNA Quantitation Assay Kit can be used for selectively detecting as little as 25 pg/ml of dsDNA in the presence of ssDNA, RNA, and free nucleotides.
- High sensitivity
- Exceptional DNA affinity
- Robust protocol
- Multiple applications
Caution: For research use only.