Portelite™ Fluorimetric DNA Quantitation Kit with Broad Dynamic Range *Optimized for Cytocite™ and Qubit™ Fluorometers*
Catalog # AATB17665
Supplier: AAT Bioquest
CAS Number:
Specifications
- Bal.:100 Tests
- Popis:Portelite™ fluorimetric DNA quantitation kit with broad dynamic range *optimized for cytocite™ and qubit™ fluorometers*
Specifications
About this item
DNA Quantitation is a very important task in DNA sample preparations for various analyses.
- Linear over three orders of magnitude
- Exceptional dsDNA selectivity over rna
- Robust protocol
- Multiple applications
Caution: For research use only.