Lumiwox™ Acridinium NHS Ester
Katalogové číslo AATB26000
Dodavatel: AAT BIOQUEST
Číslo CAS:
Specifikace
- Bal.:1 mg
- Popis:Lumiwox™ acridinium NHS ester
Specifikace
O tomto produktu
The acridinium NHS ester can be used to label proteins and nucleic acids.
- Chemiluminescent labeling
- Improved sensitivity and stronger signal
- Tolerates hydrogen peroxide reaction
- Suitable for various experimental surfaces
- Multiple applications
Caution: For research use only.