ApoSight™ Green Caspase 3/7 substrate
Catalog # AATB13290
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Bal.:100 Tests
- Popis:ApoSight™ Green Caspase 3/7 substrate
Specifications
About this item
ApoSight™ Green Caspase 3/7 substrate is the first fluorogenic probe for the direct fluorescence imaing of caspase activities in live cells.
- unique fluorogenic caspase activity probe for live cells
- sensitive and accurate detection
- can be adapted for automation
Caution: Storage: Freeze (< −15 °C)