RS-232 Cable
RS-232 cable, 25-pin – 9-pin, RS-232, 3 m
Catalog # METR6.2125.110
Supplier: METROHM
Jump to:
Product Details & Documents
Connecting RS-232 interface to PC. Used with pH meters, titrinos, KF coulometers, VA trace analyser.
Specifications
- Type:RS-232
- Popis:RS-232 cable, 25-pin – 9-pin
- Délka:3 m