Orders LinkContactUsLinkComponent
Sprayer
Sprayer
Sprayer, complete
Catalog # 217-0695
Supplier:   Lenz Laborglas
Sprayer
Catalog # 217-0695
Supplier:  Lenz Laborglas
Sprayer
Sprayer
Sprayer, complete
Price
2 170,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass sprayer.
Delivery information: Complete sprayer consists of 100 ml Erlenmeyer flask with socket NS 19/26, sprayer head with clamp and rubber bulb.
Documents
Specifications
  • Popis:
    Sprayer, complete
Accessories
Product Information
  • Catalog #
    Popis
    Availability
    Price
  • 201-1434
    Sprayer head, NS 19/26, without flask or rubber bulb
    Log in for availability
    1 * 1 KS - 1 380,00 Kč
Product Information
  • Catalog #
    Popis
    Availability
    Price
  • LENZ05470005
    Rubber bulb
    Log in for availability
    1 * 1 KS - 462,00 Kč