Centrifugal concentrators, Vivacon® 2
For samples up to 2 ml; membrane: Regenerated cellulose Hydrosart®. Vivacon® 2 centrifugal concentrators have been designed for DNA concentration, buffer exchange applications and primer removal. They feature a horizontal membrane design that is preferred by some users for reasons of consistency with previously used procedures and reproducibility.
- Optimal performance with very dilute samples, example forensic specimens
- Low binding materials; high recoveries even from diluted samples
- Re-spin possibility; complete and highly reproducible sample recovery
- Volume graduation and the molecular weight cut-off printed on individual devices
- Catalog No:
- 518-0021
- 518-0019
- 518-0015
- 518-0013
- 518-0017
- Filtrační plocha:
- 0,95 cm²
- 0,95 cm²
- 0,95 cm²
- 0,95 cm²
- 0,95 cm²
- Mrtvý prostor:
- 10 µl
- 10 µl
- 10 µl
- 10 µl
- 10 µl
- Ø×H:
- 16×115/125 mm
- 16×115/125 mm
- 16×115/125 mm
- 16×115/125 mm
- 16×115/125 mm
Frequently Bought Together