0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteiny
Recombinant Human PIP (from Plasma)
Recombinant Human PIP (from Plasma)
ABNOP9063.10UG
:
Abnova
:
Recombinant Human PIP (from Plasma)
ABNOP9063.10UG
:
Abnova
:
Bal.:
10 µG
Typ proteinu/peptidu:
Recombinant
Zdroj:
Plasma
Druhy:
Human
Identifikační číslo genu:
P12273
Název proteinu/peptidu:
PIP