0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteiny
Recombinant Rat PPBP (from E. coli)
Recombinant Rat PPBP (from E. coli)
ABNOP9556.10UG
:
Abnova
:
Recombinant Rat PPBP (from E. coli)
ABNOP9556.10UG
:
Abnova
:
Bal.:
10 µG
Typ proteinu/peptidu:
Recombinant
Zdroj:
E. coli
Druhy:
Rat
Identifikační číslo genu:
Q99ME0
Název proteinu/peptidu:
PPBP
Sekvence:
46a.a.-111a.a.