ContactUsLinkComponent
Anti-GLB1 Mouse Recombinant Antibody [clone: 13R4]
Anti-GLB1 Mouse Recombinant Antibody [clone: 13R4]
Catalog # ABNORAB03691.200UG
Supplier:  Abnova
Anti-GLB1 Mouse Recombinant Antibody [clone: 13R4]
Catalog # ABNORAB03691.200UG
Supplier:  Abnova
Specifications
  • Symbol antigenu:
    GLB1
  • Klonovalita:
    Recombinant
  • Klon:
    13R4
  • Hostitel:
    Mouse
  • Izotyp:
    IgG3 lambda
  • Reaktivita:
    Human
  • Imunogen:
    Recombinant mouse monoclonal antibody raised against human GLB1.
  • Bal.:
    200 µG