ContactUsLinkComponent
Anti-MET Rabbit Recombinant Antibody [clone: R02-4D6]
Anti-MET Rabbit Recombinant Antibody [clone: R02-4D6]
Catalog # ABNORAB02107.100UL
Supplier:  Abnova
Anti-MET Rabbit Recombinant Antibody [clone: R02-4D6]
Catalog # ABNORAB02107.100UL
Supplier:  Abnova
Specifications
  • Symbol antigenu:
    MET
  • Klonovalita:
    Recombinant
  • Klon:
    R02-4D6
  • Hostitel:
    Rabbit
  • Reaktivita:
    Human
  • Imunogen:
    Recombinant rabbit monoclonal antibody raised against human MET.
  • Bal.:
    100 µl