Mobile Phase Filters
Catalog # MICS58846-05
Supplier: MICROSOLV
Specifications
- Porozita:5 µm
- Popis:Mobile phase filter
- Materiál:PTFE
- Bal.:1
- Pro zkumavky s vnějš. Ø:1,58 mm
Specifications
About this item
Biocompatible, last drop, PEEK and polypropylene mobile phase filters. PTFE filter element and will provide very high filter capacity and throughput while using a maximum amount of solvent. The unique design allows for this flat filter to vacuum up to the last drop of liquid in the reservoir bottle.
- Tubing connector uses the fitting and an ETFE ferrule to hold non-flanged low pressure tubing in the filter without leaking
- The 5 µm pore size element will filter undissolved particles from your solvents that are 5 µm or larger preventing them from damaging your HPLC pumps, columns or instrument
- ARE-applied research brand one each
The advantage of a white filter element is that they make it easy to quickly see particles that have been trapped by the filter and if the element needs to be replaced to maintain proper flow of your solvents. These filter elements are soft and easy to remove and replace.