Kits: whose DNA was left behind?
Catalog # 763-0008
Supplier: EDVOTEK
Specifications
- Popis:Whose DNA Was Left Behind?
- Bal.:1 KIT
Specifications
About this item
DNA obtained from a single hair left behind at a crime scene can be used to identify a criminal. In this experiment, students will compare simulated crime scene DNA with that of two suspects.
- For 10 lab groups
- Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Delivery: Kit includes instructions, Ready-to-Load™ dye sample, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.