Orders LinkContactUsLinkComponent
Microtest tubes
Microtest tubes
Micro test tubes, 1,5 ml
Catalog # EDVO630
Supplier:   EDVOTEK
Microtest tubes
Catalog # EDVO630
Supplier:  EDVOTEK
Microtest tubes
Micro test tubes, 1,5 ml
Price
1 300,00 Kč
Log in for availability
1 * 500 KS
Product Details & Documents
500×1,5 ml tubes.
Specifications
  • Kapacita:
    1,5 ml
  • Popis:
    Micro test tubes