Pipetting pump, electronic
Pipette pump, electronic
Katalogové číslo 613-1364
Dodavatel: EDVOTEK
Přejít na:
Product Details & Documents
This lightweight, cordless pipetting controller is ideally suited as an aliquoting tool for instructors and teaching assistants. It uses all standard serological pipettes. The speed can be fine-tuned by applying varying finger pressure to the operating buttons.
Specifikace
- Popis:Pipette pump, electronic