Experimentation kits, how does a doctor test for AIDS?
Catalog # EDVOS-70
Supplier: EDVOTEK
Specifications
- Popis:How Does a Doctor Test for AIDS?
- Bal.:1
Specifications
About this item
The body defends itself from attack by infectious agents like bacteria and viruses by producing antibodies. Enzyme linked immunosorbent assays (ELISA) test for antibodies present in the blood, which indicate infection. In this experiment, students perform a simulated ELISA test to identify infected samples and compare them to control samples.
- For 10 lab groups
- Complete in 45 minutes
Delivery: Kit includes instructions, antigens, positive and negative controls, sera, secondary antibody, substrate, detection strips, transfer pipettes, and test tubes.