Orders LinkContactUsLinkComponent
Experimentation kits, Quick PCR
Experimentation kits, Quick PCR
Quick PCR
Catalog # 763-0002
Supplier:   EDVOTEK
Experimentation kits, Quick PCR
Catalog # 763-0002
Supplier:  EDVOTEK
Experimentation kits, Quick PCR
Experimentation kits, Quick PCR
Quick PCR
Price
3 170,00 Kč
Log in for availability
1 * 1 KIT
Product Details & Documents
This experiment uses PCR to amplify a small section of Lambda DNA via a 2-step process, saving valuable classroom time and allowing completion of the lab in one session.
  • For 10 lab groups
  • Complete in 60 minutes
Also required: 5 to 50 µl adjustable micropipettes, tips, thermal cycler, electrophoresis tank, and power supply.
Delivery information: Kit includes instructions, PCR beads, DNA template and primers, DNA size ladder, ultrapure water, wax beads, gel loading dye, agarose, electrophoresis buffer, and gel stain.
Specifications
  • Popis:
    Quick PCR