Experimentation kits, PCR-based VNTR human DNA typing
Katalogové číslo EDVO334
Dodavatel: EDVOTEK
Specifikace
- Popis:PCR-based VNTR Human DNA Typing
- Bal.:1 KIT
Specifikace
O tomto produktu
In DNA fingerprinting, variable number tandem repeats (VNTR) are used to identify individuals. Using this kit, students type themselves at the D1S80 locus on chromosome 1. This region contains between 14 and 40 copies of a 16 base pair repeat.
- For 25 students
- Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Delivery: Kit includes instructions, proteinase K, PCR beads, control DNA and primers, microtubes, chelating agent, agarose, DNA ladder, practice gel loading solution, gel loading dye, electrophoresis buffer, and gel stain.