Experimentation kits, PCR-based Alu-Human DNA typing
Katalogové číslo EDVO333
Dodavatel: EDVOTEK
Specifikace
- Popis:PCR-based Alu-Human DNA Typing
- Bal.:1 KIT
Specifikace
O tomto produktu
Students use primers for a 300 base pair Alu insertion in chromosome 16 (PV92) to determine their own genotype. They can then compare their class results with others around the world over the internet.
- For 25 students
- Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Delivery: Kit includes instructions, proteinase K, PCR beads, control DNA and primers, microtubes, chelating agent, agarose, DNA ladder, practice gel loading solution, gel loading dye, electrophoresis buffer, and gel stain.