ContactUsLinkComponent
Kits: in search of the sickle cell gene by southern blot, Ready-to-Load™
Kits: in search of the sickle cell gene by southern blot, Ready-to-Load™
Katalogové číslo EDVO315
Dodavatel:  EDVOTEK
undefined
Kits: in search of the sickle cell gene by southern blot, Ready-to-Load™
Katalogové číslo EDVO315
Dodavatel:  EDVOTEK
Specifikace
  • Popis:
    In Search of the Sickle Cell Gene by Southern Blot
  • Environmentally Preferable:
  • Bal.:
    1
Specifikace
O tomto produktu
Southern blotting is an important technique used widely in clinical genetics and research. By transferring DNA from an agarose gel onto a membrane the method allows the precise analysis and identification of DNA bands on a gel. Using this kit, students use Southern blotting to find a point mutation in the haemoglobin gene indicating Sickle Cell Anaemia.
  • For five lab groups
  • Electrophoresis, 45 minutes; blotting, overnight; staining and destaining, 10 minutes
Also required: Electrophoresis tank, power supply, water bath and 80 °C incubator.
Delivery: Kit includes instructions, Ready-to-Load™ DNA samples, agarose, electrophoresis buffer, nylon membranes, filter paper, and blotting stain.