Experimentation kits, DNA/RNA microarrays
Katalogové číslo EDVO235
Dodavatel: EDVOTEK
Specifikace
- Popis:DNA/RNA Microarrays
- Bal.:1 KIT
Specifikace
O tomto produktu
Microarray technology allows scientists to screen large numbers of samples in one assay. This technology allows high-throughput screening of small samples and allows comparisons to be made between healthy and unhealthy tissues. In this experiment, students apply simulated DNA and RNA samples to a membrane to screen for positive and negative samples.
- For 10 lab groups
- Complete in 60 minutes
Also required: 5 µl fixed volume or adjustable micropipette and tips.
Informace o dodávce: Kit includes instructions, simulated patient DNA and RNA samples and controls, microarray cards, plastic bags to incubate membrane, microtest tubes and pipettes.