ContactUsLinkComponent
Kits: DNA-based screening for smallpox, Ready-to-Load™
Kits: DNA-based screening for smallpox, Ready-to-Load™
Katalogové číslo EDVO124
Dodavatel:  EDVOTEK
undefined
Kits: DNA-based screening for smallpox, Ready-to-Load™
Katalogové číslo EDVO124
Dodavatel:  EDVOTEK
Specifikace
  • Popis:
    DNA-Based Screening for Smallpox, 6 gels
  • Bal.:
    1
Specifikace
O tomto produktu
This experiment presents a bioterrorism scenario, with students examining a simulated DNA fingerprinting test for the detection of smallpox.
  • For six lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Delivery: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® DNA stain, 1 ml calibrated drop pipette, 100 ml graduated cyclinder and microtipped transfer pipettes.