Kits: principles and practice of agarose gel electrophoresis, Ready-to-Load™
Dodavatel: EDVOTEK
Certifikáty
O tomto produktu
Ready-to-Load™ kit to show how electrophoresis separates molecules on the basis of size and charge.
- For six gels
- Complete in 45 minutes
Also required: Electrophoresis tank, power supply, adjustable micropipettes (5 to 50 µl) with tips, balance and microwave or hot plate.
Delivery: Kit includes instructions, Ready-to-Load™ dye samples, UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, calibrated micropipette, 100 ml graduated cylinder, and microtipped transfer pipettes.
Frequently Bought Together