LDPE Bucket, 610/615
KAUT2000070588
: Kautex
- Kapacita:10 L
- Popis:LDPE bucket with spout
- Rozměry:193×280×300 mm
- Bal.:1
All buckets have strong walls and are very stable for a long service life.
- Thick-walled, stable industrial and laboratory bucket
- Stainless steel carrying handle covered with plastic
- Interior graduations
- Halal certified
Buckets feature a a liter scale on the inside. The laboratory buckets with spout have a steel carrying handle covered with plastic, and are suitable for food contact.