Compression Moulded Closure for Narrow Neck Bottle Series 310
Catalog # KAUT2000971704
Supplier: Kautex
Specifications
- Závit:25 mm
- Barva:Bílá
- Materiál:Melamin
- Bal.:100
- Pro láhve:250 - 1000 ml
Specifications
About this item
Compression moulded closure is made from melamine, and features a PE-foam insert.
- Leakproof