Screw Closures for Wide Neck Amber Bottle Series 303, LDPE
Catalog # KAUT2000094578
Supplier: Kautex
Specifications
- Thread:40 mm
- Colour:Jantarová
- Materiál:LDPE
- Sekvence:02
- Environmentally Preferable:
- Bal.:1
- For bottles:250 ml
Specifications
About this item
Leakproof LDPE screw closures are packaged in polyethylene bags.