0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Domů
Mikroskopy
Accessories for Digital Microscope, DVM6 M
Accessories for Digital Microscope, DVM6 M
Accessories for Digital Microscope, DVM6 M
Catalog # LEIM10450127
Supplier:
LEICA MICROSYSTEMS
Expand image
Accessories for Digital Microscope, DVM6 M
Catalog # LEIM10450127
Supplier:
LEICA MICROSYSTEMS
Specifications
Popis:
Manual IsoPro XY stage, 6×4"
Sekvence:
19
Bal.:
1
Specifications
About this item
Product Details
Product Details