Orders LinkContactUsLinkComponent
VeriFluor™ Far-Red Passive Reference Dye
VeriFluor™ Far-Red Passive Reference Dye
Dodavatel:   Biotium
VeriFluor™ Far-Red Passive Reference Dye
Dodavatel:  Biotium
Certifikáty
VeriFluor™ Far-Red Passive Reference Dye
2 Dostupných variant níže
Product Details & Documents
A far-red passive reference dye for qPCR that frees up the ROX channel for use with JUN™ probes, EvaRuby™ qPCR dye, and other orange dyes.
  • An alternative to ROX for qPCR normalization in the red channel
  • Drop-in replacement for Mustang Purple™ on most instruments
  • Tolerates DTT in qPCR reactions, unlike Mustang Purple™
  • More soluble than ROX for easier mixing and more accurate results
  • Stand-alone dye solution for use with our Forget-Me-Not™ Master Mixes or other mixes
VeriFluor™ Far-Red is a far-red passive reference dye that is used to normalize the fluorescent reporter signal during qPCR and eliminate well-to-well variation. It is an alternative to normalization with ROX passive reference dye and a direct replacement for Mustang Purple™.
Specifications for Product Family
Specifikace
Specifikace
  • Catalog No:
  • 29087-50UL
  • 29087-500UL
  • Podmínky uskladnění:
  • Store at –10...–35 °C
  • Store at –10...–35 °C
Specifikace
Product Family Options
Informace o produktu
  • Sekvence
    Ex/Em
    Bal.
    Dostupnost
    Price
  • 29087-50UL
    VeriFluor™ far-red passive reference dye, 400× in water
    VeriFluor™ far-red passive reference dye, 400× in water
    01
    640/660 nm
    50 µl
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 50 µl
    1 * 50 µl - 4 690,00 Kč
    4 690,00 Kč
    1 * 50 µl
       4 690,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    29087-50UL
    Podmínky uskladnění
    Store at –10...–35 °C
  • 29087-500UL
    VeriFluor™ far-red passive reference dye, 400× in water
    VeriFluor™ far-red passive reference dye, 400× in water
    02
    640/660 nm
    500 µl
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 500 µl
    1 * 500 µl - 30 330,00 Kč
    30 330,00 Kč
    1 * 500 µl
       30 330,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    29087-500UL
    Podmínky uskladnění
    Store at –10...–35 °C