Supelco® Ascentis® Express OH5 HPLC and UHPLC Columns
: Merck
Ascentis® Express OH5 high-speed, high-performance liquid chromatography columns are based on the highly efficient Fused-Core® particle design.
The Ascentis® Express OH5 stationary phase is a highly polar ligand that possesses 5 hydroxyl group tethered to the silica via novel proprietary linkage chemistry.
The high performance material provides a column that can be used for traditional normal phase separations using non-polar, totally organic mobile phases of for aqueous normal phase chromatography with the typical mobile phases for hydrophilic interaction liquid chromatography (HILIC) of basic, acidic, zwitterionic or neutral compounds.
: Supplied with column test certificate inside the box.
- Catalog No:
- 53772-U
- 53776-U
- 53778-U
- 53774-U
- 50350-U
- 50355-U
- 50344-U
- 50345-U
- 50343-U
- 50348-U
- 50349-U
- 50346-U
- 50347-U
- 53767-U
- 53768-U
- 53764-U
- 53766-U
- 53769-U
- 53771-U
- 50951-U
- 53781-U
- 53782-U
- 53779-U
- 53780-U
- 50313-U
- 50314-U
- 50317-U
- 50964-U
- 50965-U
- 50962-U
- 50963-U
- 50967-U
- 50968-U
- 50959-U
- 50952-U
- 50957-U
- 53755-U
- 53757-U
- 53748-U
- 53749-U
- 50339-U
- 50341-U
- 50336-U
- 50338-U
- 50321-U
- 50322-U
- 50329-U
- 50335-U
- 50327-U
- 50328-U
- 50969-U
- 50973-U
- Popis:
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- Naplnění uhlíkem:
- 3,2%
- 3,2%
- 3,2%
- 3,2%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 3,2%
- 3,2%
- 3,2%
- 3,2%
- 3,2%
- 3,2%
- 2,8%
- 3,2%
- 3,2%
- 3,2%
- 3,2%
- 2,1%
- 2,1%
- 2,1%
- 2,8%
- 2,8%
- 2,8%
- 2,8%
- 2,8%
- 2,8%
- 2,8%
- 2,8%
- 2,8%
- 3,2%
- 3,2%
- 3,2%
- 3,2%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,1%
- 2,8%
- 2,8%
- S navazanou skupinou:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Maximální tlak:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1000 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1000 bar
- 1000 bar
- Rozsah pH:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Povrchová oblast:
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 120 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 120 m²/g
- 120 m²/g
- Kód USP:
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95