Supelco® Ascentis® Express ES-Cyano HPLC and UHPLC Columns
: Merck
Ascentis® Express ES-Cyano high-speed, high-performance liquid chromatography columns are based on the highly efficient Fused-Core® particle design.
The Fused-core particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0,5 µm thick porous shell.
The sterically protected, extensively endcapped diisopropyl-cyanopropylsilane stationary phase of Ascentis Express ES-Cyano provides a stable, reversed-phase packing that can be used for basic, acidic or neutral compounds.
: Supplied with column test certificate inside the box.
- Catalog No:
- 50562-U
- 50563-U
- 50567-U
- 50568-U
- 53497-U
- 50564-U
- 50566-U
- 51734-U
- 51724-U
- 51725-U
- 51717-U
- 51729-U
- 51732-U
- 51727-U
- 51728-U
- 50557-U
- 53491-U
- 53492-U
- 50558-U
- 53486-U
- 53489-U
- 53496-U
- 51712-U
- 53494-U
- 50559-U
- 53495-U
- 50581-U
- 50583-U
- 50577-U
- 50591-U
- 50592-U
- 50585-U
- 50588-U
- 50574-U
- 50575-U
- 50569-U
- 50570-U
- 51736-U
- 51739-U
- 50593-U
- 50597-U
- 53483-U
- 53484-U
- 53481-U
- 51709-U
- 53472-U
- 53473-U
- 53468-U
- 53470-U
- 53478-U
- 53479-U
- 53475-U
- 53476-U
- Popis:
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- Naplnění uhlíkem:
- 2,5%
- 2,5%
- 2,5%
- 2,5%
- 3,5%
- 2,5%
- 2,5%
- 3,4%
- 3,4%
- 3,4%
- 3,4%
- 3,4%
- 3,4%
- 3,4%
- 3,4%
- 2,5%
- 3,5%
- 3,5%
- 2,5%
- 3,5%
- 3,5%
- 3,5%
- 3,4%
- 3,5%
- 2,5%
- 3,5%
- 2,5%
- 2,5%
- 2,5%
- 2,5%
- 2,5%
- 2,5%
- 2,5%
- 2,5%
- 2,5%
- 2,5%
- 2,5%
- 3,4%
- 3,4%
- 2,5%
- 2,5%
- 3,5%
- 3,5%
- 3,5%
- 3,4%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- S navazanou skupinou:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Maximální tlak:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1000 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1000 bar
- 1000 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1000 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- Rozsah pH:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Povrchová oblast:
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 90 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 90 m²/g
- 135 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 120 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 120 m²/g
- 120 m²/g
- 90 m²/g
- 90 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 120 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- Kód USP:
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10