About this item
Kimtech™ Polaris™ Nitrile Gloves are built for science, made and designed for use in laboratories, life science applications, and non-sterile pharmaceutical manufacturing environments.
- High-end ambidextrous powder-free nitrile gloves deliver seamless protection whenever and wherever it is needed
- Optimized nitrile formulation (acrylonitrile-butadiene) features extended chemicals and chemotherapy drugs splash protection without compromising comfort
- Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials
- Extensively tested under stringent conditions, gloves meet AQL 0,65 for water tightness which results in a low risk of pinholes
- Recyclable through The RightCycle™ Programme where available
Powder-free, not made with latex or silicone, Kimtech™ Polaris™ Nitrile Gloves are designed to keep hands comfortable and protected whilst ensuring that research applications can be carried out contamination-free. Designated as PPE Cat III according to Regulation (EU) 2016/425 and to the Regulation 2016/425 as brought into UK law and amended and are ideal for use in higher-risk applications, as well as being food contact approved.
Kimberly-Clark Professional™ personal protective equipment is only intended for occupational workplace use and must be used in compliance with all applicable health and safety laws and standards in addition to all user instructions, warnings, and limitations accompanying each product. It is essential all product user instructions and laws and regulations on the use of each product be followed in order for the product to help protect the wearer. Misuse of personal protective equipment may result in injury, sickness, or death. For correct product selection and use, individuals should seek occupational health and safety advice from a suitably competent person. This product is suitable for sale in the EU and UK.
Specifications
- Catalog No:
- 112-6562
- 112-6563
- 112-6560
- 112-6564
- 112-6561
- Obouruké/jednoruké:
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Jednorázové/znovupoužitelné:
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Latex/bez latexu:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Pudrované/nepudrované:
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Provedení:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Typ manžety:
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Tloušťka límce:
- 0,09 mil
- 0,09 mil
- 0,09 mil
- 0,09 mil
- 0,09 mil
- Tloušťka prstu:
- 0,15 mil
- 0,15 mil
- 0,15 mil
- 0,15 mil
- 0,15 mil
- Tloušťka drapáku:
- 0,12 mil
- 0,12 mil
- 0,12 mil
- 0,12 mil
- 0,12 mil
- Regulované dle FDA:
- +
- +
- +
- +
- +
- Tažná síla:
- ≥ 14 Mpa
- ≥ 14 Mpa
- ≥ 14 Mpa
- ≥ 14 Mpa
- ≥ 14 Mpa
- Konečné prodloužení:
- ≥ 400%, min.
- ≥ 400%, min.
- ≥ 400%, min.
- ≥ 400%, min.
- ≥ 400%, min.
- Norma EN:
- EN ISO 374-1 Type B (JKOPT)
- EN ISO 374-1 Type B (JKOPT)
- EN ISO 374-1 Type B (JKOPT)
- EN ISO 374-1 Type B (JKOPT)
- EN ISO 374-1 Type B (JKOPT)
- Schváleno pro styk s potravinami:
- +
- +
- +
- +
- +
- Čí. ASTM:
- D6978
- D6978
- D6978
- D6978
- D6978